Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145766.2 | NP_665709.2 | 5 - 6 | 544 | 68 | |
XM_006239527.2 | XP_006239589.1 | 4 - 5 | 1100 | 68 | ||
XM_006239528.2 | XP_006239590.1 | 5 - 6 | 907 | 68 | ||
XM_006239529.2 | XP_006239591.1 | 5 - 6 | 828 | 68 | ||
XM_006239530.1 | XP_006239592.1 | 5 - 6 | 528 | 68 | ||
XM_006239531.2 | XP_006239593.1 | 5 - 6 | 632 | 68 | ||
GenBank mRNA | FM031851.1 | - | 5 - 6 | 544 | 68 | |
FM044847.1 | - | 5 - 6 | 515 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |