Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238796.2 | XP_006238858.1 | 8 - 9 | 751 | 94 | |
XM_006238797.2 | XP_006238859.1 | 8 - 9 | 760 | 94 | ||
GenBank mRNA | BC158560.1 | - | 8 - 9 | 701 | 9102303 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |