Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013214.4 | NP_037346.2 | 1 - 2 | 164 | 61 | |
GenBank mRNA | BC087716.1 | - | 1 - 2 | 116 | 7316866 | 61 |
CK469690.1 | - | 189 | 61 | |||
DY317682.1 | - | 1 - 2 | 248 | 61 | ||
Y09332.1 | - | 1 - 2 | 64 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |