Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106752.1 | NP_001100222.1 | 7 - 8 | 803 | 75 | |
XM_006240422.2 | XP_006240484.1 | 9 - 10 | 891 | 75 | ||
XM_006240423.2 | XP_006240485.1 | 8 - 9 | 860 | 75 | ||
XM_006240424.2 | XP_006240486.1 | 9 - 10 | 869 | 75 | ||
XM_008764762.1 | XP_008762984.1 | 7 - 8 | 649 | 75 | ||
GenBank mRNA | BC161880.1 | - | 8 - 9 | 835 | 7126409 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |