Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109277.1 | NP_001102747.1 | 1 | 2355 | 153 | |
GenBank mRNA | BC079009.1 | - | 1 | 2333 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |