Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106754.1 | NP_001100224.1 | 2 - 3 | 699 | 72 | |
XM_008764764.1 | XP_008762986.1 | 2 - 3 | 679 | 72 | ||
XM_008764765.1 | XP_008762987.1 | 2 - 3 | 674 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |