Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191984.1 | NP_001178913.1 | 5 - 6 | 781 | 61 | |
XM_006239263.2 | XP_006239325.1 | 5 - 6 | 871 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |