Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cela3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106692.1 | NP_001100162.1 | 7 - 8 | 829 | 65 | |
XM_008764220.1 | XP_008762442.1 | 7 - 8 | 879 | 65 | ||
GenBank mRNA | FQ231865.1 | - | 7 - 8 | 809 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Protease 3' Most Rattus norvegicus 1700 Probe spans exons |