Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191576.1 | NP_001178505.1 | 2 - 3 | 580 | 76 | |
XM_006239392.1 | XP_006239454.1 | 2 - 3 | 602 | 76 | ||
XM_006239393.1 | XP_006239455.1 | 2 - 3 | 602 | 76 | ||
XM_006239395.2 | XP_006239457.1 | 2 - 3 | 383 | 76 | ||
XM_006239396.2 | XP_006239458.2 | 2 - 3 | 337 | 76 | ||
XM_006239398.2 | XP_006239460.2 | 2 - 3 | 337 | 76 | ||
XM_006239399.2 | XP_006239461.1 | 2 - 3 | 602 | 76 | ||
GenBank mRNA | FQ213337.1 | - | 2 - 3 | 309 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |