Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134549.2 | NP_001128021.2 | 10 - 11 | 2619 | 95 | |
XM_006233489.2 | XP_006233551.1 | 12 - 13 | 2903 | 95 | ||
XM_006233490.2 | XP_006233552.1 | 13 - 14 | 3493 | 95 | ||
XM_006233491.2 | XP_006233553.1 | 12 - 13 | 2900 | 95 | ||
XM_008761551.1 | XP_008759773.1 | 13 - 14 | 2932 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |