Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191849.1 | NP_001178778.1 | 1 - 2 | 975 | 67 | |
XM_006233274.2 | XP_006233336.1 | 2 - 3 | 1296 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Rattus norvegicus Probe spans exons |