Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760824.1 | XP_008759046.1 | 6982 | 108 | ||
XM_226891.8 | XP_226891.6 | 5 - 6 | 1504 | 108 | ||
GenBank mRNA | D26496.1 | - | 102 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |