Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106419.1 | NP_001099889.1 | 3 - 4 | 618 | 167 | |
XM_006232058.2 | XP_006232120.1 | 3 - 4 | 536 | 167 | ||
XM_006232059.2 | XP_006232121.1 | 3 - 4 | 500 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |