Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133300.3 | NP_579834.2 | 1 - 2 | 63 | 141 | |
GenBank mRNA | DY309526.1 | - | 1 - 2 | 63 | 141 | |
FQ212901.1 | - | 1 - 2 | 49 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |