Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp21a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057101.2 | NP_476442.2 | 6 - 7 | 741 | 85 | |
XM_006255907.2 | XP_006255969.1 | 6 - 7 | 815 | 85 | ||
GenBank mRNA | U56853.1 | - | 6 - 7 | 742 | 85 | |
U56854.1 | - | 5 - 6 | 803 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |