Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc14a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019346.2 | NP_062219.2 | 2 - 3 | 262 | 75 | |
XM_008772168.1 | XP_008770390.1 | 3 - 4 | 480 | 75 | ||
XM_008772169.1 | XP_008770391.1 | 2 - 3 | 1394 | 75 | ||
XM_008772170.1 | XP_008770392.1 | 2 - 3 | 331 | 75 | ||
GenBank mRNA | X98399.1 | - | 2 - 3 | 262 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |