Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ011608.1 | - | 4 - 5 | 445 | 61 | |
BC088272.1 | - | 4 - 5 | 452 | 7379940 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |