Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031086.2 | NP_112348.2 | 11 - 12 | 1409 | 109 | |
XM_006240719.2 | XP_006240781.1 | 9 - 10 | 1695 | 109 | ||
XM_008765045.1 | XP_008763267.1 | 9 - 10 | 1141 | 109 | ||
GenBank mRNA | CV105055.1 | - | 2 - 3 | 364 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |