Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khsrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133602.1 | NP_598286.1 | 6 - 7 | 545 | 77 | |
XM_006244292.2 | XP_006244354.1 | 6 - 7 | 683 | 77 | ||
GenBank mRNA | AF308818.1 | - | 6 - 7 | 545 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |