Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037496.1 | NP_001032585.1 | 3 - 4 | 666 | 61 | |
XM_006256737.2 | XP_006256799.1 | 3 - 4 | 1316 | 61 | ||
XM_006256738.2 | XP_006256800.1 | 3 - 4 | 1315 | 61 | ||
XM_006256739.2 | XP_006256801.1 | 3 - 4 | 1314 | 61 | ||
XM_006256740.2 | XP_006256802.1 | 4 - 5 | 1186 | 61 | ||
XM_006256741.2 | XP_006256803.1 | 4 - 5 | 1185 | 61 | ||
GenBank mRNA | BC110054.1 | - | 3 - 4 | 666 | 7446041 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |