Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199115.2 | NP_954546.1 | 2 - 3 | 564 | 96 | |
GenBank mRNA | AY393999.1 | - | 2 - 3 | 424 | 96 | |
BC078944.1 | - | 2 - 3 | 564 | 7109431 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |