Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009176.2 | NP_001009176.1 | 3 - 4 | 1109 | 89 | |
GenBank mRNA | BC129078.1 | - | 3 - 4 | 1109 | 8371876 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |