Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109883.2 | NP_001103353.1 | 18 - 19 | 1979 | 65 | |
GenBank mRNA | AF293383.1 | - | 17 - 18 | 1875 | 65 | |
CV114142.1 | - | 3 - 4 | 409 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |