Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019381.2 | NP_062254.2 | 8 - 9 | 695 | 97 | |
XM_006257323.2 | XP_006257385.1 | 8 - 9 | 837 | 97 | ||
XM_008765670.1 | XP_008763892.1 | 9 - 10 | 795 | 97 | ||
GenBank mRNA | AY321320.1 | - | 10 - 11 | 833 | 97 | |
AY325130.1 | - | 10 - 11 | 833 | 97 | ||
AY325243.1 | - | 10 - 11 | 833 | 97 | ||
BC058478.1 | - | 8 - 9 | 701 | 6888036 | 97 | |
FQ217052.1 | - | 8 - 9 | 694 | 97 | ||
FQ218664.1 | - | 8 - 9 | 697 | 97 | ||
FQ226858.1 | - | 8 - 9 | 724 | 97 | ||
FQ229652.1 | - | 8 - 9 | 696 | 97 | ||
FQ230608.1 | - | 8 - 9 | 724 | 97 | ||
FQ231817.1 | - | 8 - 9 | 702 | 97 | ||
X75855.1 | - | 8 - 9 | 724 | 97 | ||
X75856.1 | - | 7 - 8 | 608 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |