Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005545.1 | NP_001005545.1 | 4 - 5 | 643 | 63 | |
GenBank mRNA | BC082112.1 | - | 4 - 5 | 643 | 7135321 | 63 |
FQ213994.1 | - | 2 - 3 | 708 | 63 | ||
FQ230067.1 | - | 4 - 5 | 673 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |