Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rorc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232926.2 | XP_006232988.2 | 2 - 3 | 155 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Nuclear Receptor Rattus norvegicus Probe spans exons |