Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108071.1 | NP_001101541.1 | 5 - 6 | 507 | 109 | |
XM_006240915.2 | XP_006240977.1 | 7 - 8 | 1436 | 109 | ||
XM_006240916.2 | XP_006240978.1 | 7 - 8 | 1426 | 109 | ||
XM_006240917.2 | XP_006240979.1 | 7 - 8 | 1433 | 109 | ||
XM_006240918.2 | XP_006240980.1 | 7 - 8 | 1376 | 109 | ||
XM_006240919.2 | XP_006240981.1 | 7 - 8 | 1435 | 109 | ||
XM_006240920.2 | XP_006240982.1 | 6 - 7 | 1265 | 109 | ||
GenBank mRNA | FQ224249.1 | - | 7 - 8 | 736 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |