Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb41l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021681.2 | NP_067713.2 | 22 - 23 | 4606 | 150 | |
NM_172090.3 | NP_742087.1 | 19 - 20 | 2573 | 150 | ||
XM_008762381.1 | XP_008760603.1 | 22 - 23 | 4606 | 150 | ||
GenBank mRNA | AB019256.1 | - | 19 - 20 | 2571 | 150 | |
AB019257.1 | - | 22 - 23 | 4606 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |