Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108599.1 | NP_001102069.1 | 1 - 2 | 210 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |