Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017279.1 | NP_058975.1 | 3 - 4 | 257 | 72 | |
GenBank mRNA | BC060576.1 | - | 3 - 4 | 276 | 6919255 | 72 |
FQ214776.1 | - | 3 - 4 | 272 | 72 | ||
FQ223872.1 | - | 272 | 72 | |||
J02897.1 | - | 3 - 4 | 257 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |