Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017279.1 | NP_058975.1 | 6 - 7 | 532 | 129 | |
GenBank mRNA | BC060576.1 | - | 6 - 7 | 551 | 6919255 | 129 |
FQ214776.1 | - | 6 - 7 | 547 | 129 | ||
FQ223872.1 | - | 547 | 129 | |||
J02897.1 | - | 6 - 7 | 532 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |