Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199099.2 | NP_954530.1 | 4 - 5 | 373 | 76 | |
XM_006243751.2 | XP_006243813.1 | 4 - 5 | 414 | 76 | ||
GenBank mRNA | BC060585.1 | - | 4 - 5 | 374 | 6891123 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |