Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013004.1 | NP_037136.1 | 19 - 20 | 2446 | 79 | |
GenBank mRNA | AJ001637.1 | - | 19 - 20 | 2446 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |