Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zwint | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147138.2 | NP_671479.1 | 4 - 5 | 513 | 73 | |
GenBank mRNA | AF439397.1 | - | 4 - 5 | 495 | 73 | |
AF532776.1 | - | 4 - 5 | 513 | 73 | ||
BC063144.1 | - | 4 - 5 | 505 | 6921645 | 73 | |
EU000468.1 | - | 4 - 5 | 481 | 73 | ||
FQ228814.1 | - | 2 - 3 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |