Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012514.1 | NP_036646.1 | 13 - 14 | 4475 | 124 | |
XM_008768092.1 | XP_008766314.1 | 13 - 14 | 4691 | 124 | ||
GenBank mRNA | AF036760.1 | - | 13 - 14 | 4475 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |