Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031799.1 | NP_113987.1 | 1 - 2 | 63 | 83 | |
XM_006236231.2 | XP_006236293.1 | 2 - 3 | 6116 | 83 | ||
XM_006236232.2 | XP_006236294.1 | 3 - 4 | 1112 | 83 | ||
XM_006236233.2 | XP_006236295.1 | 3 - 4 | 554 | 83 | ||
GenBank mRNA | AF053100.1 | - | 1 - 2 | 63 | 83 | |
AF053101.1 | - | 1 - 2 | 63 | 83 | ||
AF053102.1 | - | 1 - 2 | 63 | 83 | ||
AF053103.1 | - | 1 - 2 | 63 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |