Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198761.1 | NP_942056.1 | 4 - 5 | 1678 | 119 | |
GenBank mRNA | AY382879.1 | - | 4 - 5 | 1678 | 119 | |
FQ228283.1 | - | 2071 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |