Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201562.3 | NP_963856.1 | 6 - 7 | 1158 | 64 | |
GenBank mRNA | AY279211.1 | - | 6 - 7 | 1158 | 64 | |
BC129079.1 | - | 6 - 7 | 1273 | 8370402 | 64 | |
FQ215784.1 | - | 3 - 4 | 410 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |