Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ooep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766421.1 | XP_008764643.1 | 2 - 3 | 437 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |