Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234464.2 | XP_006234526.1 | 1 - 2 | 116 | 87 | |
GenBank mRNA | FQ216695.1 | - | 132 | 87 | ||
FQ217513.1 | - | 1 - 2 | 77 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |