Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013936.1 | NP_001013958.1 | 4 - 5 | 881 | 71 | |
GenBank mRNA | BC083723.1 | - | 4 - 5 | 881 | 7191003 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |