Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134556.1 | NP_001128028.1 | 6 - 7 | 817 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |