Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105819.1 | NP_001099289.1 | 2 - 3 | 388 | 83 | |
XM_006246982.2 | XP_006247044.1 | 2 - 3 | 450 | 83 | ||
XM_008767963.1 | XP_008766185.1 | 2 - 3 | 450 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |