Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270598.1 | NP_001257527.1 | 13 - 14 | 1522 | 97 | |
NM_001270599.1 | NP_001257528.1 | 13 - 14 | 1522 | 97 | ||
GenBank mRNA | FQ222567.1 | - | 13 - 14 | 1290 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |