Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015021.1 | NP_001015021.1 | 3 - 4 | 510 | 130 | |
GenBank mRNA | AY387070.1 | - | 3 - 4 | 330 | 130 | |
BC093384.1 | - | 3 - 4 | 510 | 7389370 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |