Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751518.3 | XP_003751566.1 | 3 - 4 | 824 | 108 | |
XM_006252367.2 | XP_006252429.1 | 3 - 4 | 862 | 108 | ||
XM_006252368.2 | XP_006252430.1 | 2 - 3 | 714 | 108 | ||
XM_006252369.2 | XP_006252431.1 | 3 - 4 | 824 | 108 | ||
XM_008770927.1 | XP_008769149.1 | 3 - 4 | 553 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |