Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770082.1 | XP_008768304.1 | 1 - 2 | 272 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |