Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106883.1 | NP_001100353.1 | 2 - 3 | 820 | 71 | |
GenBank mRNA | BC168177.1 | - | 2 - 3 | 807 | 9035251 | 71 |
FQ214885.1 | - | 2 - 3 | 822 | 71 | ||
FQ214919.1 | - | 2 - 3 | 712 | 71 | ||
FQ215260.1 | - | 2 - 3 | 822 | 71 | ||
FQ216053.1 | - | 2 - 3 | 822 | 71 | ||
FQ216174.1 | - | 2 - 3 | 712 | 71 | ||
FQ216808.1 | - | 2 - 3 | 708 | 71 | ||
FQ217810.1 | - | 2 - 3 | 822 | 71 | ||
FQ224967.1 | - | 2 - 3 | 855 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |