Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106554.1 | NP_001100024.1 | 5 - 6 | 648 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |