Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115042.2 | NP_001108514.2 | 1 - 2 | 153 | 80 | |
XM_008760176.1 | XP_008758398.1 | 1 - 2 | 148 | 80 | ||
GenBank mRNA | BC097411.1 | - | 1 - 2 | 83 | 80 | |
BC158663.1 | - | 1 - 2 | 153 | 8369770 | 80 | |
BC158764.1 | - | 1 - 2 | 92 | 7111047 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |